Learn More
Meridian Life Science 1-Step qPCR Buffer 4x, 50ML
SDP
Catalog No. 5000856424
Encompass_Preferred
1-Step qPCR Buffer, 4x is a novel buffer system that has been designed for highly reproducible, accurate reverse transcription followed by qPCR in a single tube under fast thermal cycling conditions, delivering excellent results in fast RT-qPCR assays.
Catalog No. 50-008-56424 Supplier Meridian Life Science Supplier No. MDX03450
1-Step qPCR Buffer, 4x is a novel buffer system that has been designed for highly reproducible, accurate reverse transcription followed by qPCR in a single tube under fast thermal cycling conditions, delivering excellent results in fast RT-qPCR assays.