Learn More
Embi Tec GE-3750: 1% Agarose TAE w/EtBr Landscape Gel 24+1 Well
SDP
Catalog No. NC1013869
Encompass
1% Agarose TAE w/EtBr Landscape Gel 24+1 WellEasy to UseJust open gel package and place gel with tray directly onto the RunOne Unit platform or similiar size platform.ConvenientKeep stock on hand with a 7 Month Shelf Life. Store at 4-24° C.
Catalog No. NC1013869 Supplier Embi Tec Supplier No. GG3750
1% Agarose TAE w/EtBr Landscape Gel 24+1 WellEasy to UseJust open gel package and place gel with tray directly onto the RunOne Unit platform or similiar size platform.ConvenientKeep stock on hand with a 7 Month Shelf Life. Store at 4-24° C.