Learn More
Medchemexpress LLC 1-(tert-Butoxycarbonyl)-2-methylpyrrolidine-2-carboxylic acid | 203869-80-1 | 99.3% | C11H19NO4 | 1 G
SDP
Catalog No. 5000434510
Encompass_Preferred
1-(tert-Butoxycarbonyl)-2-methylpyrrolidine-2-carboxylic acid (Boc-α-me-dl-pro-oh) is a drug intermediate used for the synthesis of various active compounds.
  • Appears as a solid
  • Color is white to off-white
  • Store as powder at -20°C for 3 years, or 4°C for 2 years
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month
Catalog No. 50-004-34510 Supplier Medchemexpress LLC Supplier No. HY761361G
1-(tert-Butoxycarbonyl)-2-methylpyrrolidine-2-carboxylic acid (Boc-α-me-dl-pro-oh) is a drug intermediate used for the synthesis of various active compounds.
  • Appears as a solid
  • Color is white to off-white
  • Store as powder at -20°C for 3 years, or 4°C for 2 years
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month