Learn More
Medchemexpress LLC 1-(tert-Butoxycarbonyl)-2-methylpyrrolidine-2-carboxylic acid | 203869-80-1 | 99.3% | C11H19NO4 | 250 MG
SDP
Catalog No. 5000358605
Encompass_Preferred
1-(tert-Butoxycarbonyl)-2-methylpyrrolidine-2-carboxylic acid is a drug intermediate for the synthesis of various active compounds.
  • Acts as a drug intermediate
  • Used for synthesis of various active compounds
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-003-58605 Supplier Medchemexpress LLC Supplier No. 250MGHY76136
1-(tert-Butoxycarbonyl)-2-methylpyrrolidine-2-carboxylic acid is a drug intermediate for the synthesis of various active compounds.
  • Acts as a drug intermediate
  • Used for synthesis of various active compounds
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month