Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(tert-Butoxycarbonyl)-2-methylpyrrolidine-2-carboxylic acid | 203869-80-1 | MFCD00223594 | 99.3% | C11H19NO4 | 500 MG
SDP

Catalog No. 5000434173
Encompass_Preferred

1-(tert-Butoxycarbonyl)-2-methylpyrrolidine-2-carboxylic acid, also known as Boc-α-me-dl-pro-oh, is a solid, white to off-white compound primarily used as a drug intermediate in the synthesis of various active compounds. It is intended for research use only.

  • Used as a drug intermediate
  • Synthesizes various active compounds
  • Appears as a white to off-white solid
  • Purity of 99.25%
  • Molecular weight of 229.28
  • Chemical formula C11H19NO4
  • CAS number 203869-80-1

Catalog No. 50-004-34173 Supplier Medchemexpress LLC Supplier No. HY76136500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

1-(tert-Butoxycarbonyl)-2-methylpyrrolidine-2-carboxylic acid, also known as Boc-α-me-dl-pro-oh, is a solid, white to off-white compound primarily used as a drug intermediate in the synthesis of various active compounds. It is intended for research use only.

  • Used as a drug intermediate
  • Synthesizes various active compounds
  • Appears as a white to off-white solid
  • Purity of 99.25%
  • Molecular weight of 229.28
  • Chemical formula C11H19NO4
  • CAS number 203869-80-1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.