Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(tert-Butoxycarbonyl)-3-fluoro-4-piperidone 97.0+%, TCI America™
SDP

Catalog No. B4858200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4858200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4858200MG Supplier TCI America Supplier No. B4858200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 211108-50-8
Molecular Formula C10H16FNO3
Molecular Weight (g/mol) 217.24
MDL Number MFCD10566554
InChI Key JZNWQLLPLOQGOI-UHFFFAOYSA-N
Synonym 1-boc-3-fluoro-4-oxopiperidine, 3-fluoro-4-oxopiperidine-1-carboxylic acid tert-butyl ester, 1-boc-3-fluoro-4-piperidone, 1-tert-butoxycarbonyl-3-fluoro-4-piperidone, 1-piperidinecarboxylic acid, 3-fluoro-4-oxo-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-fluoro-4-oxopiperidine, 3-fluoro-4-oxo-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl3-fluoro-4-oxopiperidine-1-carboxylate, tert-butyl 3-fluoro-4-oxo-piperidine-1-carboxylate, n-boc-3-fluoro-4-piperidinone
PubChem CID 10560711
IUPAC Name tert-butyl 3-fluoro-4-oxopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(=O)C(C1)F

Specifications

Melting Point 77°C
Color White-Yellow
Quantity 200 mg
Formula Weight 217.24
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(tert-Butoxycarbonyl)-3-fluoro-4-piperidone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.