Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(tert-Butoxycarbonyl)-3-piperidone 98.0+%, TCI America™
SDP

Catalog No. B33095G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33095G 5 g
B330925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33095G Supplier TCI America Supplier No. B33095G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 98977-36-7
Molecular Formula C10H17NO3
Molecular Weight (g/mol) 199.25
MDL Number MFCD01631193
InChI Key RIFXIGDBUBXKEI-UHFFFAOYSA-N
Synonym 1-boc-3-piperidone, 1-boc-3-piperidinone, n-boc-3-piperidone, 1-n-boc-3-piperidone, n-boc-3-piperidinone, boc-3-piperidone, 1-tert-butoxycarbonyl-3-piperidone, 3-oxo-piperidine-1-carboxylic acid tert-butyl ester, 3-oxopiperidine, n-boc protected, tert-butyl 3-oxopiperidinecarboxylate
PubChem CID 2756825
IUPAC Name tert-butyl 3-oxopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(=O)C1

Specifications

Melting Point 38°C
Color White-Yellow
Quantity 5 g
Formula Weight 199.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-(tert-Butoxycarbonyl)-3-piperidone
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.