Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-tert-Butoxycarbonyl-4-iodopiperidine 97.0+%, TCI America™
SDP

Catalog No. B41645G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41645G 5 g
B41641G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41645G Supplier TCI America Supplier No. B41645G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 301673-14-3
Molecular Formula C10H18INO2
Molecular Weight (g/mol) 311.163
MDL Number MFCD04115041
InChI Key YFWQFKUQVJNPKP-UHFFFAOYSA-N
Synonym n-boc-4-iodopiperidine, 1-boc-4-iodopiperidine, n-boc-4-iodo-piperidine, 1-tert-butoxycarbonyl-4-iodopiperidine, tert-butyl 4-iodotetrahydro-1 2h-pyridinecarboxylate, 1-n-boc-4-iodopiperidine, 4-iodo-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl 4-iodopiperidinecarboxylate, tert-butyl 4-iodo-1-piperidinecarboxylate
PubChem CID 10892302
IUPAC Name tert-butyl 4-iodopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)I

Specifications

Melting Point 46°C
Color White-Yellow
Quantity 5 g
Formula Weight 311.16
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-tert-Butoxycarbonyl-4-iodopiperidine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.