Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-[(tert-Butoxycarbonyl)amino]cyclopropanecarboxylic acid, ≥97%, Thermo Scientific™

Catalog No. SB02070DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SB02070DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SB02070DA Supplier Thermo Scientific Chemicals Supplier No. SB02070DA

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 88950-64-5
Molecular Formula C9H15NO4
Molecular Weight (g/mol) 201.22
MDL Number MFCD00083257
InChI Key DSKCOVBHIFAJRI-UHFFFAOYSA-N
Synonym 1-boc-amino cyclopropanecarboxylic acid, 1-tert-butoxycarbonyl amino cyclopropanecarboxylic acid, n-boc-1-aminocyclopropanecarboxylic acid, 1-tert-butoxycarbonylamino cyclopropanecarboxylic acid, boc-acpc-oh, 1-tert-butoxy carbonyl amino cyclopropane-1-carboxylic acid, 1-boc amino cyclopanecarboxylic acid, 1-tert-butoxycarbonyl amino cyclopropane-1-carboxylic acid, cyclopropanecarboxylic acid, 1-1,1-dimethylethoxy carbonyl amino
PubChem CID 688396
IUPAC Name 1-{[(tert-butoxy)carbonyl]amino}cyclopropane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1(CC1)C(O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 201.22
Percent Purity 97%
Chemical Name or Material 1-[(Tert-butoxycarbonyl)amino]cyclopropanecarboxylic acid
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.