Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(tert-Butoxycarbonyl)piperidine 98.0+%, TCI America™
SDP

Catalog No. B46255G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46255G 5 g
B462525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46255G Supplier TCI America Supplier No. B46255G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 75844-69-8
Molecular Formula C10H19NO2
Molecular Weight (g/mol) 185.267
MDL Number MFCD02093935
InChI Key RQCNHUCCQJMSRG-UHFFFAOYSA-N
Synonym 1-boc-piperidine, n-boc-piperidine, 1-tert-butoxycarbonyl piperidine, 1-piperidinecarboxylic acid, 1,1-dimethylethyl ester, piperidine-1-carboxylic acid tert-butyl ester, tert-butyl piperidinecarboxylate, boc-piperidine, n-boc piperidine, pubchem7258, n-boc tetrahydropyridine
PubChem CID 7010304
IUPAC Name tert-butyl piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCCC1

Specifications

Color Colorless
Boiling Point 120°C
Quantity 5 g
Formula Weight 185.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(tert-Butoxycarbonyl)piperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.