Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(tert-Butoxycarbonylamino)cyclopentanecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. B38311G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B38311G Supplier TCI America Supplier No. B38311G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 35264-09-6
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.276
MDL Number MFCD01076126
InChI Key YBZCSKVLXBOFSL-UHFFFAOYSA-N
Synonym boc-cycloleucine, 1-boc-amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1-n-boc-aminocyclopentanecarboxylic acid, boc-1-aminocyclopentane-1-carboxylic acid, n-boc-cycloleucine, 1-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, 1-tert-butoxy carbonyl amino cyclopentane-1-carboxylic acid
PubChem CID 2734645
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1(CCCC1)C(=O)O

Specifications

Melting Point 133°C
Color White
Quantity 1 g
Formula Weight 229.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(tert-Butoxycarbonylamino)cyclopentanecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.