Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-tert-Butyl-3-ethoxybenzene | 133073-81-1 | 95.0% | C12H18O | 1 G
SDP

Catalog No. 5000434523
Encompass_Preferred

1-tert-Butyl-3-ethoxybenzene is a chemical compound that serves as a drug intermediate, crucial for the synthesis of various active pharmaceutical compounds.

  • Appearance: liquid, light yellow to light brown
  • Purity: 95.0%
  • Molecular weight: 178.28
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-34523 Supplier Medchemexpress LLC Supplier No. HY759811G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-tert-Butyl-3-ethoxybenzene is a chemical compound that serves as a drug intermediate, crucial for the synthesis of various active pharmaceutical compounds.

  • Appearance: liquid, light yellow to light brown
  • Purity: 95.0%
  • Molecular weight: 178.28
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.