Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-tert-Butyl-3-ethoxybenzene | 133073-81-1 | 95.0% | C12H18O | 250 MG
SDP

Catalog No. 5000358377
Encompass_Preferred

1-tert-Butyl-3-ethoxybenzene is a drug intermediate used for the synthesis of various active compounds. It is for research use only.

  • Molecular weight: 178.28
  • Appearance: Liquid
  • Color: Light yellow to light brown

Catalog No. 50-003-58377 Supplier Medchemexpress LLC Supplier No. 250MGHY75981
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-tert-Butyl-3-ethoxybenzene is a drug intermediate used for the synthesis of various active compounds. It is for research use only.

  • Molecular weight: 178.28
  • Appearance: Liquid
  • Color: Light yellow to light brown

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.