Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-tert-Butyl-3-ethoxybenzene | 133073-81-1 | 95.0% | C12H18O | 5 G
SDP

Catalog No. 5000434660
Encompass_Preferred

1-tert-Butyl-3-ethoxybenzene is a drug intermediate used for the synthesis of various active compounds. It has a molecular weight of 178.28 and an appearance of light yellow to light brown liquid.

  • Used for the synthesis of various active compounds
  • Molecular weight: 178.28
  • Appearance: Light yellow to light brown liquid

Catalog No. 50-004-34660 Supplier Medchemexpress LLC Supplier No. HY759815G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-tert-Butyl-3-ethoxybenzene is a drug intermediate used for the synthesis of various active compounds. It has a molecular weight of 178.28 and an appearance of light yellow to light brown liquid.

  • Used for the synthesis of various active compounds
  • Molecular weight: 178.28
  • Appearance: Light yellow to light brown liquid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.