Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-tert-Butyl-3-ethylbenzene, 98%

Catalog No. AAB2032506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2032506 5 g
AAB2032514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2032506 Supplier Thermo Scientific Chemicals Supplier No. B2032506
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 14411-56-4 | C12H18 | 162.276 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14411-56-4
Molecular Formula C12H18
Molecular Weight (g/mol) 162.276
MDL Number MFCD00060845
InChI Key MUJPTTGNHRHIPH-UHFFFAOYSA-N
PubChem CID 139753
IUPAC Name 1-tert-butyl-3-ethylbenzene
SMILES CCC1=CC(=CC=C1)C(C)(C)C

Specifications

Density 0.86
Boiling Point 205°C
Odor Mild
Quantity 5 g
Synonym 1-t-butyl-3-ethylbenzene, 1-tert-butyl-3-ethylbenzene, benzene, 1-1,1-dimethylethyl-3-ethyl, benzene,1-1,1-dimethylethyl-3-ethyl, butylethylbenzene, m-tert-butyl-ethylbenzene, acmc-1cb13, 1-tert-butyl-3-ethyl-benzene, 1-1,1-dimethylethyl-3-ethylbenzene, 1-tert-butyl-3-ethylbenzene 5g
Formula Weight 162.27
Percent Purity 98%
Chemical Name or Material 1-tert-Butyl-3-ethylbenzene
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.