Learn More
Medchemexpress LLC 1-tert-butyl 3-methyl 3-hydroxyazetidine-1,3-dicarboxylate | 1884303-05-2 | MFCD30530458 | 95.0% | 231.25 g/mol | C10H17NO5 | 250 MG
SDP
Catalog No. 5000209663
Encompass_Preferred
1-tert-butyl 3-methyl 3-hydroxyazetidine-1,3-dicarboxylate is a protected azetidine-based synthetic intermediate used in medicinal chemistry and organic synthesis as a building block for active compound synthesis. It is supplied in small-scale research quantities and requires cold storage for long-term stability.
  • Protected azetidine intermediate for multi-step synthesis.
  • Suitable as a drug intermediate for synthesis of active compounds.
  • Reported purity 95.0% by NMR.
  • Molecular formula C10H17NO5; molecular weight 231.25 g/mol.
  • Store pure material at -20°C for long-term stability.
Catalog No. 50-002-09663 Supplier Medchemexpress LLC Supplier No. 250MGHYW066651
1-tert-butyl 3-methyl 3-hydroxyazetidine-1,3-dicarboxylate is a protected azetidine-based synthetic intermediate used in medicinal chemistry and organic synthesis as a building block for active compound synthesis. It is supplied in small-scale research quantities and requires cold storage for long-term stability.
  • Protected azetidine intermediate for multi-step synthesis.
  • Suitable as a drug intermediate for synthesis of active compounds.
  • Reported purity 95.0% by NMR.
  • Molecular formula C10H17NO5; molecular weight 231.25 g/mol.
  • Store pure material at -20°C for long-term stability.