Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-tert-Butyl-6-iodo-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one

Catalog No. ADE0012671G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ADE0012671G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ADE0012671G Supplier Sigma Aldrich Supplier No. ADE0012671G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2,5-diaminopyridine is a disubstituted pyridine that can be prepared by the reduction of 5-nitro-2-aminopyridine using 10%pd/c.2,5-diaminopyridine may be used in the preparation of 6-acetamidoimidazo[ 1,2-a]pyridine
  • It may be used in the preparation of organo-soluble polyimides by reacting with different dianhydride monomers
  • These polyimides form the precursor for preparing novel polyimide-silica composites

Specifications

CAS 1203499-26-6
Molecular Formula C11H13IN2O2
Linear Formula C11H13IN2O2
MDL Number MFCD13563081
Quantity 10 g
Molecular Weight (g/mol) 332.14
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.