Learn More
Medchemexpress LLC 1-tert-butyl N-(5R)-substituted triazole-4-carboxamide (BIIB091) | 2247614-80-6 | 99.9% | 542.64 g/mol | C28H34N10O2 | 5 MG
SDP
Catalog No. 5000221744
Encompass_Preferred
BIIB091 is a potent, selective, orally active, reversible Bruton tyrosine kinase (BTK) inhibitor supplied for laboratory research. It shows sub-nanomolar enzymatic potency and is provided as solid material or as a DMSO solution for in vitro and preclinical studies; refer to the manufacturer datasheet for safety, handling, and the certificate of analysis.
  • Sub-nanomolar BTK potency (IC50 <0.5 nM).
  • Reversible and highly selective BTK inhibition.
  • Chemical formula C28H34N10O2; molecular weight 542.64 g/mol.
  • Available as solid quantities and as a 10 mM solution in DMSO.
  • High purity (>99.8% by HPLC) suitable for research applications.
Catalog No. 50-002-21744 Supplier Medchemexpress LLC Supplier No. HY1399845MG
BIIB091 is a potent, selective, orally active, reversible Bruton tyrosine kinase (BTK) inhibitor supplied for laboratory research. It shows sub-nanomolar enzymatic potency and is provided as solid material or as a DMSO solution for in vitro and preclinical studies; refer to the manufacturer datasheet for safety, handling, and the certificate of analysis.
  • Sub-nanomolar BTK potency (IC50 <0.5 nM).
  • Reversible and highly selective BTK inhibition.
  • Chemical formula C28H34N10O2; molecular weight 542.64 g/mol.
  • Available as solid quantities and as a 10 mM solution in DMSO.
  • High purity (>99.8% by HPLC) suitable for research applications.