Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Tetradecanethiol

Catalog No. 871935ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
87-193-5ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 87-193-5ML Supplier Sigma Aldrich Supplier No. 871935ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Tetradecanethiol (TDT) is an alkanethiol used as a self-assembled monolayer (SAM) which may be used to functionalize a variety of nanoparticles. It is a hydrophobic ligand. It is highly oriented and acts as a hole-injection barrier on the nanoparticle surface.TDT is used as a self-assembled monolayer (SAM) on gold nanoparticle(AuNP) based electrodes for bi-enzymer glucose biosensors. The surface modification was done by soaking the electrodes in TDT solution.

Specifications

CAS 2079-95-0
Density 0.846 g/mL (at 20°C)
Molecular Formula C14H30S
Refractive Index n20/D 1.461
Linear Formula CH3(CH2)13 SH
Quantity 5 mL
MDL Number MFCD00022099
Molecular Weight (g/mol) 230.45
Percent Purity ≥98.0% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.