Learn More
Medchemexpress LLC 1-Tetradecanol | 112-72-1 | 214.39 | 1 KG
SDP
Supplier:  Medchemexpress LLC HYW0042941KG
Encompass_Preferred
1-Tetradecanol, also known as Myristyl Alcohol, is a straight-chain saturated fatty alcohol. This compound, which can be isolated from *Myristica fragrans*, functions as an antibacterial agent. It is recognized for its various biological activities, including its role as an antibacterial and anti-inflammatory agent, particularly in applications related to periodontitis.
  • Straight-chain saturated fatty alcohol
  • Isolated from Myristica fragrans
  • Exhibits antibacterial activity
  • Possesses anti-inflammatory properties
  • Used in research for periodontitis
Catalog No. 50-004-45963