Learn More
Medchemexpress LLC 1-Tetradecanol | 112-72-1 | 214.39 | 1 ML
SDP
Catalog No. 5000447675
Encompass_Preferred
1-Tetradecanol is a straight-chain saturated fatty alcohol that acts as an antibacterial agent. It can be isolated from Myristica fragrans. This compound exhibits both antibacterial and anti-inflammatory activities, particularly in the context of periodontitis.
  • Reduces IL-8 production by AGS cells in response to H. pylori in a dose-dependent manner.
  • Significantly reduces melanin in B16F10 cells dose-dependently without causing cell cytotoxicity.
Catalog No. 50-004-47675 Supplier Medchemexpress LLC Supplier No. HYW00429410MM/1ML
1-Tetradecanol is a straight-chain saturated fatty alcohol that acts as an antibacterial agent. It can be isolated from Myristica fragrans. This compound exhibits both antibacterial and anti-inflammatory activities, particularly in the context of periodontitis.
  • Reduces IL-8 production by AGS cells in response to H. pylori in a dose-dependent manner.
  • Significantly reduces melanin in B16F10 cells dose-dependently without causing cell cytotoxicity.