Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Tetradecanol | 112-72-1 | 214.39 | 100 G
SDP

Catalog No. 5000446962
Encompass_Preferred

1-Tetradecanol is a straight-chain saturated fatty alcohol known for its antibacterial and anti-inflammatory properties. It can be isolated from Myristica fragrans. Studies indicate that it can reduce IL-8 production by AGS cells in response to H. pylori and significantly reduce melanin in B16F10 cells.

  • Appearance: Solid
  • Color: White to off-white
  • Formula: C14H30O
  • Initial source: Plants (other families)
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Store at room temperature for 3 years (solid); in solvent: -80°C for 2 years, -20°C for 1 year

Catalog No. 50-004-46962 Supplier Medchemexpress LLC Supplier No. HYW004294100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Tetradecanol is a straight-chain saturated fatty alcohol known for its antibacterial and anti-inflammatory properties. It can be isolated from Myristica fragrans. Studies indicate that it can reduce IL-8 production by AGS cells in response to H. pylori and significantly reduce melanin in B16F10 cells.

  • Appearance: Solid
  • Color: White to off-white
  • Formula: C14H30O
  • Initial source: Plants (other families)
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Store at room temperature for 3 years (solid); in solvent: -80°C for 2 years, -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.