Learn More
Medchemexpress LLC 1-Tetradecanol | 112-72-1 | 214.39 | 25 G
SDP
Catalog No. 5000449473
Encompass_Preferred
1-Tetradecanol is a straight-chain saturated fatty alcohol and an antibacterial agent. It is isolated from Myristica fragrans and exhibits antibacterial and anti-inflammatory activity, particularly against periodontitis.
  • Reduces IL-8 production by AGS cells in response to H. pylori
  • Significantly reduces melanin in B16F10 cells without cytotoxicity
Catalog No. 50-004-49473 Supplier Medchemexpress LLC Supplier No. 25GHYW004294
1-Tetradecanol is a straight-chain saturated fatty alcohol and an antibacterial agent. It is isolated from Myristica fragrans and exhibits antibacterial and anti-inflammatory activity, particularly against periodontitis.
  • Reduces IL-8 production by AGS cells in response to H. pylori
  • Significantly reduces melanin in B16F10 cells without cytotoxicity