Learn More
Medchemexpress LLC 1-Tetradecanol | 112-72-1 | 214.39 | 50 G
SDP
Catalog No. 5000445962
Encompass_Preferred
1-Tetradecanol is a straight-chain saturated fatty alcohol that also acts as an antibacterial agent. It can be isolated from Myristica fragrans and has shown both antibacterial and anti-inflammatory activity, particularly in relation to periodontitis.
  • Straight-chain saturated fatty alcohol
  • Antibacterial agent
  • Anti-inflammatory activity (periodontitis)
  • Reduces IL-8 production in AGS cells in response to H. pylori
  • Reduces melanin in B16F10 cells without cytotoxicity
Catalog No. 50-004-45962 Supplier Medchemexpress LLC Supplier No. HYW00429450G
1-Tetradecanol is a straight-chain saturated fatty alcohol that also acts as an antibacterial agent. It can be isolated from Myristica fragrans and has shown both antibacterial and anti-inflammatory activity, particularly in relation to periodontitis.
  • Straight-chain saturated fatty alcohol
  • Antibacterial agent
  • Anti-inflammatory activity (periodontitis)
  • Reduces IL-8 production in AGS cells in response to H. pylori
  • Reduces melanin in B16F10 cells without cytotoxicity