Learn More
Medchemexpress LLC 1-Tetradecanol-d2 | 169398-02-1 | 216.40 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW004294S1100MG
Encompass_Preferred
1-Tetradecanol-d2 is a deuterium-labeled version of 1-Tetradecanol, a straight-chain saturated fatty alcohol originally isolated from Myristica fragrans. This compound is known for its antibacterial and anti-inflammatory properties, particularly in treating periodontitis. Deuteration, the process of incorporating stable heavy isotopes of hydrogen, can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-46961