Learn More
Medchemexpress LLC 1-Tetradecanol-d2 | 169398-02-1 | 216.40 | 25 MG
SDP
Catalog No. 5000446831
Encompass_Preferred
1-Tetradecanol-d2 is the deuterium labeled 1-Tetradecanol, a straight-chain saturated fatty alcohol isolated from Myristica fragrans. This compound possesses antibacterial and anti-inflammatory (periodontitis) activity.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs
  • Isolated from Myristica fragrans
  • Possesses antibacterial and anti-inflammatory activity
Catalog No. 50-004-46831 Supplier Medchemexpress LLC Supplier No. 25MGHYW004294S1
Add to Cart
Add to Cart
1-Tetradecanol-d2 is the deuterium labeled 1-Tetradecanol, a straight-chain saturated fatty alcohol isolated from Myristica fragrans. This compound possesses antibacterial and anti-inflammatory (periodontitis) activity.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs
  • Isolated from Myristica fragrans
  • Possesses antibacterial and anti-inflammatory activity