Learn More
Medchemexpress LLC 1-Tetradecanol-d2 | 169398-02-1 | 216.40 | 50 MG
SDP
Catalog No. 5000448534
Encompass_Preferred
1-Tetradecanol-d2 is the deuterium-labeled derivative of 1-Tetradecanol, a straight-chain saturated fatty alcohol isolated from Myristica fragrans. This compound is known for its antibacterial and anti-inflammatory activities, particularly against periodontitis. It is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs due to deuterium substitution.
Catalog No. 50-004-48534 Supplier Medchemexpress LLC Supplier No. HYW004294S150MG
1-Tetradecanol-d2 is the deuterium-labeled derivative of 1-Tetradecanol, a straight-chain saturated fatty alcohol isolated from Myristica fragrans. This compound is known for its antibacterial and anti-inflammatory activities, particularly against periodontitis. It is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs due to deuterium substitution.