Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Tetradecene

Catalog No. T98051L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T98051L 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T98051L Supplier Sigma Aldrich Supplier No. T98051L
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Octyn-3-ol is a racemic intermediate formed during the synthesis of enantiomerically pure secondary alcohols with sterically similar substituents.1-Octyn-3-ol was used in the synthesis of synthetic tricolorin A, a novel tetrasaccharide macrolactone that is a natural herbicide .

Specifications

CAS 1120-36-1
Melting Point -13°C to +11°C (lit.)
Density 0.775 g/mL (at 25°C (literature))
Boiling Point 251°C (lit.)
Molecular Formula C14H28
Refractive Index n20/D 1.436 (literature)
Linear Formula CH3(CH2)11 CH=CH2
MDL Number MFCD00008981
Quantity 250 g
Molecular Weight (g/mol) 196.37
Percent Purity 92%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.