Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Tetradecene 99.5+%, TCI America™
SDP

Catalog No. S03445ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03445ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03445ML Supplier TCI America Supplier No. S03445ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1120-36-1
Molecular Formula C14H28
Molecular Weight (g/mol) 196.378
MDL Number MFCD00008981
InChI Key HFDVRLIODXPAHB-UHFFFAOYSA-N
Synonym 1-tetradecene, n-tetradec-1-ene, tetradecene, 1-tetradecylene, alpha-tetradecene, unii-fw23481s7s, ccris 3785, alkenes, c10-16 .alpha., a-tetradecene, tetradecene-1
PubChem CID 14260
ChEBI CHEBI:77505
IUPAC Name tetradec-1-ene
SMILES CCCCCCCCCCCCC=C

Specifications

Color Colorless
Quantity 5 mL
Formula Weight 196.38
Percent Purity ≥99.5% (GC)
Physical Form Liquid
Chemical Name or Material 1-Tetradecene [Standard Material for GC]
EINECSNumber (2)-0027
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.