Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1-Tetradecyl)triphenylphosphonium bromide, 97%

Catalog No. AAL0431122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0431122 100 g
AAL0431114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0431122 Supplier Thermo Scientific Chemicals Supplier No. L0431122
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 25791-20-2 | C32H44BrP | 539.58 g/mol

(1-Tetradecyl)triphenylphosphonium bromide is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Tetradecyl)triphenylphosphonium bromide is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Slightly soluble in Water.

Notes
Hygroscopic. Store away from oxidizing agents. Incompatible with moisture and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25791-20-2
Molecular Formula C32H44BrP
Molecular Weight (g/mol) 539.58
MDL Number MFCD00051857
InChI Key FUMBGFNGBMYHGH-UHFFFAOYSA-M
Synonym triphenyl tetradecyl phosphonium bromide, myristyltriphenylphosphonium bromide, 1-tetradecyl triphenylphosphonium bromide, triphenyl tetradecyl phosphanium bromide, tetradecyl triphenylphosphonium bromide, acmc-209gmk, tetradecyltriphenylphosphoniumbromide, triphenyl tetradecyl phosphoniumbromide, n-tetradecyltriphenylphosphonium bromide, phosphonium,triphenyltetradecyl-, bromide 1:1
PubChem CID 3084442
IUPAC Name triphenyl(tetradecyl)phosphanium;bromide
SMILES [Br-].CCCCCCCCCCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 87°C to 89°C
Quantity 100 g
Beilstein 3582243
Sensitivity Hygroscopic
Solubility Information Slightly soluble in Water.
Formula Weight 539.59
Percent Purity 97%
Chemical Name or Material (1-Tetradecyl)triphenylphosphonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 247-267-1
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.