Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Tetradecylboronic acid, Thermo Scientific™

Catalog No. AAL1996614
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1996614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1996614 Supplier Thermo Scientific Chemicals Supplier No. L1996614

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100888-40-2
Molecular Formula C14H31BO2
Molecular Weight (g/mol) 242.21
MDL Number MFCD00093676
InChI Key MCUAMUGSKSODIJ-UHFFFAOYSA-N
Synonym n-tetradecylboronic acid, boronic acid,b-tetradecyl, tetradecylboranic acid, acmc-20eg95, tetradecaneboronic acid
PubChem CID 3528229
IUPAC Name tetradecylboronic acid
SMILES CCCCCCCCCCCCCCB(O)O

Specifications

Melting Point 72°C to 76°C
Quantity 25 g
Beilstein 1757986
Formula Weight 242.21
Chemical Name or Material 1-Tetradecylboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.