Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Tetrahydro-2H-pyran-4-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 98.0+%, TCI America™
SDP

Catalog No. T36681G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T36681G 1 g
T3668200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T36681G Supplier TCI America Supplier No. T36681G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1040377-03-4
Molecular Formula C14H23BN2O3
Molecular Weight (g/mol) 278.159
MDL Number MFCD12033229
InChI Key BOOVIFJKQGYEON-UHFFFAOYSA-N
Synonym 1-(Tetrahydro-2H-pyran-4-yl)-1H-pyrazole-4-boronic Acid Pinacol Ester
PubChem CID 45788496
IUPAC Name 1-(oxan-4-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)C3CCOCC3

Specifications

Melting Point 115°C
Color White-Yellow
Quantity 1 g
Formula Weight 278.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(Tetrahydro-2H-pyran-4-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.