Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(Tetrahydropyran-2-yl)-1H-pyrazole-5-boronic acid pinacol ester | 903550-26-5 | 278.16 | 5 G
SDP

Catalog No. 5000446964
Encompass_Preferred

This biochemical reagent is suitable as a biological material or organic compound for life science related research. It is intended for research use only and not for sale to patients.

  • Appearance: Solid, white to yellow
  • Formula: C14H23BN2O3
  • Melting point: 74-78°C
  • Boiling point: 413.8±35.0 °C at 760 mmHg
  • Solubility (in vitro): DMSO: 100 mg/mL (359.50 mM)
  • Storage (as supplied): -20°C, sealed, away from moisture
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month (sealed, away from moisture)
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Identified uses: Laboratory chemicals, manufacture of substances
  • Hazards: Acute toxicity (oral), skin/eye/respiratory irritation

Catalog No. 50-004-46964 Supplier Medchemexpress LLC Supplier No. HYW0012425G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This biochemical reagent is suitable as a biological material or organic compound for life science related research. It is intended for research use only and not for sale to patients.

  • Appearance: Solid, white to yellow
  • Formula: C14H23BN2O3
  • Melting point: 74-78°C
  • Boiling point: 413.8±35.0 °C at 760 mmHg
  • Solubility (in vitro): DMSO: 100 mg/mL (359.50 mM)
  • Storage (as supplied): -20°C, sealed, away from moisture
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month (sealed, away from moisture)
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Identified uses: Laboratory chemicals, manufacture of substances
  • Hazards: Acute toxicity (oral), skin/eye/respiratory irritation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.