Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-thiazol-2-yl-piperazine

Catalog No. CDS004812 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS004812 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS004812 Supplier Sigma Aldrich Supplier No. CDS004812100MG

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Chlorosulfonylacetyl chloride has been used in:
  • Preparation of novel tricyclic benzothiazolo [2,3-c] thiadiazine antagonists of the platelet adp receptor
  • N-terminal derivatization of peptides in rapid determination of peptide or protein sequences by matrix-assisted laser desorption ionization mass spectrometry

Specifications

CAS 42270-37-1
Molecular Formula C7H11N3S
Linear Formula C7H11N3S
MDL Number MFCD00778419
Quantity 1 g
Molecular Weight (g/mol) 169.25
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.