Learn More
Medchemexpress LLC 1-Thio-B-D-glucose s 100mg | 10593-29-0 | 218.20 | C6H11NaO5S | 100 MG
SDP
Catalog No. 5000228751
Encompass_Preferred
1-Thio-β-D-glucose sodium is the sodium salt form of 1-thio-β-D-glucose supplied as a solid reagent for biochemical and surface chemistry applications. It is used to form hydrophilic monolayers on metals and to stabilize lipid bilayers and proteins.
  • Sodium salt form of 1-thio-β-D-glucose
  • Forms hydrophilic self-assembled monolayers on metal surfaces
  • Stabilizes lipid bilayers and helps protect proteins from denaturation
  • High aqueous solubility (≥ 100 mg/mL) and soluble in DMSO (≈ 50 mg/mL)
  • Available in small laboratory quantities, including 100 MG packs
  • Powder storage: -20°C for long-term stability; in solvent store at -80°C for extended storage
Catalog No. 50-002-28751 Supplier Medchemexpress LLC Supplier No. HY115419100MG
1-Thio-β-D-glucose sodium is the sodium salt form of 1-thio-β-D-glucose supplied as a solid reagent for biochemical and surface chemistry applications. It is used to form hydrophilic monolayers on metals and to stabilize lipid bilayers and proteins.
  • Sodium salt form of 1-thio-β-D-glucose
  • Forms hydrophilic self-assembled monolayers on metal surfaces
  • Stabilizes lipid bilayers and helps protect proteins from denaturation
  • High aqueous solubility (≥ 100 mg/mL) and soluble in DMSO (≈ 50 mg/mL)
  • Available in small laboratory quantities, including 100 MG packs
  • Powder storage: -20°C for long-term stability; in solvent store at -80°C for extended storage