Learn More
Medchemexpress LLC 1-Thio-B-D-glucose s 25mg | 10593-29-0 | 25 MG
SDP
Catalog No. 5000227095
Encompass_Preferred
1-Thio-β-D-glucose sodium is the sodium salt of 1-thio-β-D-glucose used as a biochemical reagent. It forms hydrophilic self-assembled monolayers on metal surfaces, stabilizes lipid bilayers, and can protect proteins from denaturation. Supplied as a solid powder for laboratory research and typically stored at low temperatures for long-term stability.
  • Sodium salt form of 1-thio-β-D-glucose.
  • Solid powder suitable for laboratory research.
  • Forms hydrophilic self-assembled monolayers with metal surfaces.
  • Stabilizes lipid bilayers and protects proteins from denaturation.
  • Suitable for small-scale mg quantities and low-temperature storage.
Catalog No. 50-002-27095 Supplier Medchemexpress LLC Supplier No. HY11541925MG
1-Thio-β-D-glucose sodium is the sodium salt of 1-thio-β-D-glucose used as a biochemical reagent. It forms hydrophilic self-assembled monolayers on metal surfaces, stabilizes lipid bilayers, and can protect proteins from denaturation. Supplied as a solid powder for laboratory research and typically stored at low temperatures for long-term stability.
  • Sodium salt form of 1-thio-β-D-glucose.
  • Solid powder suitable for laboratory research.
  • Forms hydrophilic self-assembled monolayers with metal surfaces.
  • Stabilizes lipid bilayers and protects proteins from denaturation.
  • Suitable for small-scale mg quantities and low-temperature storage.