Learn More
Medchemexpress LLC 1-Thio-B-D-glucose s 50mg | 10593-29-0 | 50 MG
SDP
Catalog No. 5000228752
Encompass_Preferred
1-Thio-β-D-glucose sodium is the sodium salt form of 1-thio-β-D-glucose supplied as a solid reagent for biochemical and surface chemistry research. It forms hydrophilic self-assembled monolayers on metal surfaces, stabilizes lipid bilayers, and can protect proteins from denaturation. Available in small milligram pack sizes for laboratory use.
  • Forms hydrophilic self-assembled monolayers with metals.
  • Stabilizes lipid bilayer and protects proteins from denaturation.
  • Provided as a solid powder with defined storage conditions: powder -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).
  • Available in small pack sizes suitable for research (25 mg-500 mg).
Catalog No. 50-002-28752 Supplier Medchemexpress LLC Supplier No. HY11541950MG
1-Thio-β-D-glucose sodium is the sodium salt form of 1-thio-β-D-glucose supplied as a solid reagent for biochemical and surface chemistry research. It forms hydrophilic self-assembled monolayers on metal surfaces, stabilizes lipid bilayers, and can protect proteins from denaturation. Available in small milligram pack sizes for laboratory use.
  • Forms hydrophilic self-assembled monolayers with metals.
  • Stabilizes lipid bilayer and protects proteins from denaturation.
  • Provided as a solid powder with defined storage conditions: powder -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).
  • Available in small pack sizes suitable for research (25 mg-500 mg).