Learn More
Cayman Chemical 1-thio-β-D-Glucose Tetraacetate, Purity: ≥95%, CAS: 19879-84-6, 1 g
SDP
Catalog No. 502992078
Encompass_Preferred
A building block; has been used in the synthesis of aromatic glucosinolates with anti-inflammatory activity, as well as glucosylated poly(pentafluorostyrene) derivatives for coating magnetic iron oxide nanoparticles
Catalog No. 50-299-2078 Supplier Cayman Chemical Supplier No. 304461 G
A building block; has been used in the synthesis of aromatic glucosinolates with anti-inflammatory activity, as well as glucosylated poly(pentafluorostyrene) derivatives for coating magnetic iron oxide nanoparticles