Learn More
Cayman Chemical 1-thio-β-D-Glucose (sodium salt), Purity: ≥95%, CAS: 10593-29-0, 500 mg
SDP
Catalog No. 502970479
Encompass_Preferred
An analog of β-D-glucose in which sulfur replaces the hydroxyl group at the one position, allowing diverse chemical reactions, including click chemistry and polymerization; can be labeled with technetium-99m for analytical and diagnostic procedures
Catalog No. 50-297-0479 Supplier Cayman Chemical Supplier No. 16485500 MG
An analog of β-D-glucose in which sulfur replaces the hydroxyl group at the one position, allowing diverse chemical reactions, including click chemistry and polymerization; can be labeled with technetium-99m for analytical and diagnostic procedures