Learn More
Cayman Chemical 1-ThIoglycerol 10mg
SDP
Catalog No. 502985168
Encompass_Preferred
A reagent used in fluorescence, spectroscopy, and microbiology techniques; provides precise control in varying the shell size of ultrasmall quantum dots; used as a capping agent in spectroscopy; stimulates porphyrin synthesis and increases glutamyl-tRNA reductase activity in E. coli grown in an aerobic environment; previously used in hair permanents
Catalog No. 50-298-5168 Supplier Cayman Chemical Supplier No. 2082410 MG
A reagent used in fluorescence, spectroscopy, and microbiology techniques; provides precise control in varying the shell size of ultrasmall quantum dots; used as a capping agent in spectroscopy; stimulates porphyrin synthesis and increases glutamyl-tRNA reductase activity in E. coli grown in an aerobic environment; previously used in hair permanents