Learn More
1-Thionaphthol, 99%
Description
1-Thionaphthol is used to produce the di-[1]naphthyl disulfide. It is also used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Thionaphthol is used to produce the di-[1]naphthyl disulfide. It is also used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in diethyl ether and ethanol. Slightly soluble in water and dilute alkalis.
Notes
Store away from air. Store under dry inert gas. Incompatible with air and oxidizing agents.
Specifications
Specifications
| Quantity | 25 g |
| Sensitivity | Air Sensitive |
| Solubility Information | Soluble in diethyl ether and ethanol. Slightly soluble in water and dilute alkalis. |
| Formula Weight | 160.24 |
| Percent Purity | 99% |
| Chemical Name or Material | 1-Thionaphthol |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement H301-H315-H319-H335 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement P261-P280-P301+P310-P305+P351+P338-P405-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| EINECSNumber | 208-462-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.