Learn More
Origen Biomedical 1 TO 1 SPIKE PORT 12CM TUBING
Catalog No. NC1211933
Encompass
  • Sterile, single-use design for safe fluid transfer
  • 12 cm tubing length for efficient reach
  • Compatible with CryoStore™ bags for flexible usage
  • One spike to one spike port and two female luers configuration
  • Integrated pinch clamps for controlled fluid management
  • Individually pouched for easy storage and handling
  • Case quantity of 20 units for convenient bulk supply
Catalog No. NC1211933 Supplier Origen Biomedical Supplier No. SFFP12
  • Sterile, single-use design for safe fluid transfer
  • 12 cm tubing length for efficient reach
  • Compatible with CryoStore™ bags for flexible usage
  • One spike to one spike port and two female luers configuration
  • Integrated pinch clamps for controlled fluid management
  • Individually pouched for easy storage and handling
  • Case quantity of 20 units for convenient bulk supply