Learn More
Thermco Products Inc Precision lab Hg thermometer, Fractional degree, -1 deg./201 deg.C, 0.1 deg.C div, TOTAL imm, 610mm
Catalog No. NC0596319
Encompass_Preferred
High precision mercury SAMA thermometer that meet or exceed the specifications of the American National Standards Institute. Specifications approve by N.I.S.T. and ASTM. Individually serialized and permanetly marked. Made with precision yellow backed tubing. SAMA No. CT50
Catalog No. NC0596319 Supplier Thermco Products Inc Supplier No. ACC1007
High precision mercury SAMA thermometer that meet or exceed the specifications of the American National Standards Institute. Specifications approve by N.I.S.T. and ASTM. Individually serialized and permanetly marked. Made with precision yellow backed tubing. SAMA No. CT50