Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Tosylimidazole | 2232-08-8 | MFCD00005285 | 222.27 | 250 G
SDP

Catalog No. 5000449778
Encompass_Preferred

1-Tosylimidazole is a biochemical reagent widely utilized in life science research. It functions as both a biological material and an organic compound, essential for various laboratory applications.

  • Appears as a white to off-white solid
  • Purity greater than 99.75%
  • Molecular formula C10H10N2O2S
  • CAS number 2232-08-8
  • Solubility in DMSO is 100 mg/mL

Catalog No. 50-004-49778 Supplier Medchemexpress LLC Supplier No. 250GHYW001965
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Tosylimidazole is a biochemical reagent widely utilized in life science research. It functions as both a biological material and an organic compound, essential for various laboratory applications.

  • Appears as a white to off-white solid
  • Purity greater than 99.75%
  • Molecular formula C10H10N2O2S
  • CAS number 2232-08-8
  • Solubility in DMSO is 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.