Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Triacontanol 85.0+%, TCI America™
SDP

Catalog No. T10491G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10491G 1 g
T1049100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T10491G Supplier TCI America Supplier No. T10491G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 593-50-0
Molecular Formula C30H62O
Molecular Weight (g/mol) 438.825
MDL Number MFCD00002963
InChI Key REZQBEBOWJAQKS-UHFFFAOYSA-N
Synonym 1-triacontanol, triacontanol, melissyl alcohol, myricyl alcohol, n-triacontanol, triacontyl alcohol, triacontanol-1, miraculan, myricyl alcohol van, 1-hydroxytriacontane
PubChem CID 68972
ChEBI CHEBI:28409
IUPAC Name triacontan-1-ol
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCO

Specifications

Melting Point 86°C
Quantity 1 g
Formula Weight 438.83
Percent Purity ≥85.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Triacontanol
EINECSNumber (2)-0217
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.