Learn More
Medchemexpress LLC 1-(triazolo[1,5-a]pyridin-2-yl)-N-((1S)-1-(2-fluoro-4-(trifluoromethyl)phenyl)ethyl)cyclopropanecarb | 2654741-64-5 | 98.2% | 431.45 g/mol | C22H24F3N5O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1449625MG
Encompass_Preferred
SOS1-IN-11 is a small-molecule SOS1 inhibitor for preclinical research. It inhibits SOS1 with an IC50 of 30 nM and is supplied as a light yellow to yellow solid with high purity, suitable for biochemical and cell-based studies.
  • Potent SOS1 inhibitor (IC50 30 nM).
  • High purity (98.2%).
  • Molecular weight 431.45 g/mol.
  • Supplied as 5 mg solid for small-scale studies.
  • Powder stable at -20°C for up to 3 years.
  • Suitable for biochemical and cell-based SOS1 assays.
Catalog No. 50-002-21514