Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Tridecanol 97%, Thermo Scientific™

Catalog No. AC139530500
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC139530500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC139530500 Supplier Thermo Scientific Chemicals Supplier No. 139530500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-70-9
Molecular Formula C13H28O
Molecular Weight (g/mol) 200.366
InChI Key XFRVVPUIAFSTFO-UHFFFAOYSA-N
Synonym 1-tridecanol, tridecanol, tridecyl alcohol, n-tridecan-1-ol, n-tridecanol, n-tridecyl alcohol, alcohols, c12-14, ccris 8591, dsstox_cid_1947, dsstox_rid_76420
PubChem CID 8207
ChEBI CHEBI:34123
IUPAC Name tridecan-1-ol
SMILES CCCCCCCCCCCCCO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 30.5°C to 33.5°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula CH3(CH2)12OH
Quantity 50 g
Beilstein 01, 428
Formula Weight 200.36
Percent Purity ≥96.0%
Physical Form Low Melting Solid
Chemical Name or Material 1-Tridecanol, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.