Learn More
Medchemexpress LLC 1-Tridecanol-d27 | 203633-18-5 | 227.53 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW004293S5MG
Encompass_Preferred
1-Tridecanol-d27 is a deuterium-labeled variant of 1-Tridecanol. This compound features stable heavy isotopes, making it valuable for use as tracers in drug development for quantitative analysis. Deuteration studies highlight its potential to influence the pharmacokinetic and metabolic profiles of pharmaceutical compounds.
  • Ideal for laboratory chemicals and substance manufacturing.
  • Incorporates stable heavy isotopes for quantitation.
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs.
  • Stable under recommended storage conditions.
  • For research use only by experienced personnel.
  • Appearance: white to off-white solid-liquid mixture.
  • Isotopic enrichment of 99.3%.
  • 98.7% purity (GC).
Catalog No. 50-004-48536