Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Trimethoxysilyl)naphthalene 98.0+%, TCI America™
SDP

Catalog No. T32925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32925G 5 g
T32921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T32925G Supplier TCI America Supplier No. T32925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18052-76-1
Molecular Formula C13H16O3Si
Molecular Weight (g/mol) 248.353
MDL Number MFCD00094090
InChI Key ZSOVVFMGSCDMIF-UHFFFAOYSA-N
Synonym Trimethoxy(1-naphthyl)silane
PubChem CID 11954884
IUPAC Name trimethoxy(naphthalen-1-yl)silane
SMILES CO[Si](C1=CC=CC2=CC=CC=C21)(OC)OC

Specifications

Melting Point 39°C
Color White
Boiling Point 148°C
Quantity 5 g
Formula Weight 248.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-(Trimethoxysilyl)naphthalene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.