Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-(Trimethylsiloxy)-1,3-butadiene

Catalog No. 22722650G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-722-650G 50 g
22-722-610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-722-650G Supplier Sigma Aldrich Supplier No. 22722650G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Mechanism of Diels-Alder reaction between 1-(trimethylsiloxy)-1,3-butadiene and 4,6-dinitrobenzofuroxan has been studied using state-of-the-art computational methods.1-(Trimethylsiloxy)-1,3-butadiene is commonly employed as diene in the Diels-Alder reaction.

Specifications

CAS 6651-43-0
Density 0.811 g/mL (at 25°C (literature))
Boiling Point 131°C (lit.)
Molecular Formula C7H14OSi
Refractive Index n20/D 1.448 (literature)
Linear Formula (CH3)3 SiOCH=CHCH=CH2
MDL Number MFCD00009647
Quantity 50 g
Molecular Weight (g/mol) 142.27
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.